4-Bromo-1H-indazole 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Bromoindazole
A116723-100G
A116723-10G
A116723-1G
A116723-25G
A116723-500G
A116723-5G
103824-654EA
11
USD
103824-654
77009-216
77009-218
77009-220
77009-222
77009-224
77311-448
4-Bromo-1H-indazole 97%
4-Bromo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂ MW: 197.04 g/mol Boiling Pt: 333.8 °C (760 mm Hg) Melting Pt: 165…167 °C Flash Pt: 155.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664001 CAS Number: 186407-74-9 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...