3-Amino-4-bromo-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1H-indazol-3-amine

A671283-1G A671283-5G A671283-100MG A671283-250MG
103840-498EA 154 USD
103840-498 76844-700 76844-702 77280-634 77280-636
3-Amino-4-bromo-1H-indazole 95%
3-Amino-4-bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Boiling Pt: 431.3 °C (760 mm Hg)
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD09909580
CAS Number: 914311-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR