4-Bromo-1H-benzo[d]imidazol-2-amine 97%

Supplier: Ambeed
Warning

A210638-10G A210638-250MG A210638-1G A210638-5G
103818-030EA 75 USD
103818-030 103848-724 76825-880 76825-882 77087-108 77087-110
4-Bromo-1H-benzo[d]imidazol-2-amine 97%
4-Bromo-1H-benzo[d]imidazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907621
CAS Number: 1266114-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR