4-Bromo-1-chloroisoquinoline 97%

Supplier: Ambeed
Warning

A267751-1G A267751-5G A267751-250MG A267751-25G
103812-878EA 17 USD
103812-878 103852-778 77092-526 77092-528 77262-374 77262-376 77304-988
4-Bromo-1-chloroisoquinoline 97%
4-Bromo-1-chloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Melting Pt: 93…97 °C
Flash Pt: 152.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234469
CAS Number: 66728-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR