5-Bromo-2-chloroiodobenzene 95%

Supplier: Ambeed
Warning

A211049-10G A211049-1G A211049-5G A211049-100G
103818-072EA 11 USD
103818-072 103848-766 76975-556 77087-172 77087-174 77671-884
5-Bromo-2-chloroiodobenzene 95%
5-Bromo-2-chloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClI
Storage Temperature: Refrigerator
MDL Number: MFCD07780669
CAS Number: 774608-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR