4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene

A184470-100G A184470-10G A184470-1G A184470-500G A184470-25G A184470-5G
103846-316EA 7 USD
103846-316 76769-668 76769-670 76769-672 76769-674 77083-928 77083-930
4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether 97%
4-(5-Bromo-2-chlorobenzyl)phenyl ethyl ether

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrClO
MW: 325.64 g/mol
Boiling Pt: 393 °C (760 mm Hg)
Flash Pt: 191.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11042292
CAS Number: 461432-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR