4-Bromo-1,3-thiazole-2-carboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-Bromothiazole-2-carboxylic acid

A563320-1G A563320-100MG A563320-250MG
103828-758EA 27 USD
103828-758 103839-892 76844-000 77034-928 77602-388
4-Bromo-1,3-thiazole-2-carboxylic acid 96%
4-Bromo-1,3-thiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂BrNO₂S
MW: 208.033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701413
CAS Number: 88982-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR