4-Bromo-1,3-dimethyl-1H-pyrazole 95%
Supplier: Ambeed
|
Warning
|
A130017-100G
A130017-10G
A130017-250MG
A130017-1G
A130017-25G
A130017-5G
103801-368EA
8
USD
103801-368
77027-000
77027-002
77027-004
77313-796
77313-798
77313-800
4-Bromo-1,3-dimethyl-1H-pyrazole 95%
4-Bromo-1,3-dimethyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₅H₇BrN₂ MW: 175.0265 g/mol Boiling Pt: 210.265 °C (760 mm Hg) Flash Pt: 80.966 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00985488 CAS Number: 5775-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...