4-Bromo-1,2-dihydropyridazine-3,6-dione 96%

Supplier: Ambeed
Warning

A266491-10G A266491-250MG A266491-1G A266491-5G
103812-808EA 15 USD
103812-808 103852-710 76816-118 76816-120 77092-416 77092-418
4-Bromo-1,2-dihydropyridazine-3,6-dione 96%
4-Bromo-1,2-dihydropyridazine-3,6-dione

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MW: 190.9800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923098
CAS Number: 15456-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR