4-Bromo-1,2-bis(trifluoromethyl)benzene 97%

Supplier: Ambeed
Warning

A190590-250MG A190590-1G
103816-312EA 72 USD
103816-312 103846-912 77164-266 77302-228
4-Bromo-1,2-bis(trifluoromethyl)benzene 97%
4-Bromo-1,2-bis(trifluoromethyl)benzene

Precaución: Sealed in dry, at room temperature.

Formula: C₈H₃BrF₆
MW: 293.004 g/mol
Boiling Pt: 181.6 °C (760 mm Hg)
Flash Pt: 63.7 °C
Storage Temperature: Ambient
MDL Number: MFCD06253758
CAS Number: 320-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR