4-Bromo-1-(tert-butyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A348528-1G A348528-10G
103825-994EA 6 USD
103825-994 103836-984 76840-060 77883-954
4-Bromo-1-(tert-butyl)-1H-pyrazole 95%
4-Bromo-1-(tert-butyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₁₁BrN₂
Boiling Pt: 229.8±13.0 °C (760 mm Hg)
Flash Pt: 92.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13195703
CAS Number: 70951-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR