4-Bromo-4'-hydroxybiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4'-Bromo-[1,1'-biphenyl]-4-ol

A100652-10G A100652-25G A100652-100G
103821-244EA 9 USD
103821-244 76765-116 76765-118 77570-064
4-Bromo-4'-hydroxybiphenyl 97%
4-Bromo-4'-hydroxybiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Boiling Pt: 355.5 °C (760 mm Hg)
Melting Pt: 163…168 °C
Flash Pt: 168.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00059076
CAS Number: 29558-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR