tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Boc-4,7-diazaspiro[2.5]octane

A221123-100MG A221123-10G A221123-250MG A221123-1G A221123-25G A221123-5G
103818-896EA 32 USD
103818-896 103849-776 76975-830 76975-832 76975-834 77088-370 77303-580 77303-582
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate 97%
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MW: 212.289 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08685932
CAS Number: 674792-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR