4-Boc-3-Morpholinecarboxylic acid 95%

Supplier: Ambeed
Warning

A176628-10G A176628-250MG A176628-1G A176628-25G A176628-5G
103807-156EA 15 USD
103807-156 103845-482 77011-790 77011-792 77082-832 77082-834 77082-836
4-Boc-3-Morpholinecarboxylic acid 95%
4-Boc-3-Morpholinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MW: 231.246 g/mol
Melting Pt: 38…40 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683057
CAS Number: 212650-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR