tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-2-hydroxymethylmorpholine, 4-Boc-2-Hydroxymethylmorpholine

A161560-100G A161560-10G A161560-250MG A161560-1G A161560-25G A161560-5G
103805-502EA 6 USD
103805-502 103843-740 77150-910 77150-912 77150-914 77319-726 77319-728 77319-730
tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate 98%
tert-Butyl-2-(hydroxymethyl)morpholine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Boiling Pt: 320.7 °C (760 mm Hg)
Flash Pt: 147.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426270
CAS Number: 135065-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR