4-Boc-2-(bromomethyl)-morpholine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Boc-2-(bromomethyl)morpholine
A101850-100MG
A101850-250MG
A101850-1G
A101850-5G
103796-178EA
195
USD
103796-178
103821-872
76970-184
76970-186
77308-764
77308-766
4-Boc-2-(bromomethyl)-morpholine 97%
4-Boc-2-(bromomethyl)-morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₈BrNO₃ MW: 280.159 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20489326 CAS Number: 765914-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...