4-Boc-(3-hydroxymethyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Boc-(3-Hydroxymethyl)morpholine

A196884-100MG A196884-10G A196884-250MG A196884-1G A196884-25G A196884-5G
103816-874EA 21 USD
103816-874 103847-488 76785-150 76785-152 76785-154 77085-464 77302-536 77302-538
4-Boc-(3-hydroxymethyl)morpholine 97%
4-Boc-(3-hydroxymethyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.2622 g/mol
Flash Pt: 147.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038649
CAS Number: 473923-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR