4-Biphenylacetonitrile 98%

Supplier: Ambeed
Warning

A269189-100G A269189-25G A269189-5G
103852-858EA 12 USD
103852-858 76787-148 77092-618 77305-034
4-Biphenylacetonitrile 98%
4-Biphenylacetonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00016403
CAS Number: 31603-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR