(4-Biphenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Biphenylacetic acid

A410938-25G A410938-5G A410938-100G A410938-10G A410938-1G A410938-250MG A410938-500G
103838-268EA 8 USD
103838-268 76841-796 76841-798 77278-872 77278-874 77278-876 77278-878 77278-880
(4-Biphenyl)acetic acid 98%
(4-Biphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₅C₆H₅)CH₂COOH
MW: 212.25 g/mol
Boiling Pt: 389.8 °C (760 mm Hg)
Melting Pt: 156…160 °C
Flash Pt: 286.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004351
CAS Number: 5728-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR