4-Benzyloxyindole 97%

Supplier: Ambeed
Warning

A189425-1G A189425-5G A189425-100G
103816-184EA 14 USD
103816-184 103846-776 77084-548 77084-550 77746-215
4-Benzyloxyindole 97%
4-Benzyloxyindole

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Boiling Pt: 411.6 °C (760 mm Hg)
Melting Pt: 57…61 °C
Flash Pt: 148.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00047200
CAS Number: 20289-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR