4-(Benzyloxy)-3-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A305853-100G A305853-10G A305853-25G A305853-5G
103835-616EA 9 USD
103835-616 77290-072 77290-074 77305-922 77305-924
4-(Benzyloxy)-3-methoxybenzaldehyde 98%
4-(Benzyloxy)-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Boiling Pt: 393.1 °C (760 mm Hg)
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00003365
CAS Number: 2426-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR