4-Benzylmorpholine 98%

Supplier: Ambeed
Warning

A169994-10G A169994-250MG A169994-1G A169994-25G
103806-434EA 8 USD
103806-434 103844-700 76872-372 76872-374 77301-016 77301-018
4-Benzylmorpholine 98%
4-Benzylmorpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.243 g/mol
Boiling Pt: 260.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00090503
CAS Number: 10316-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR