4-Benzoylbenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A194928-100G
A194928-10G
A194928-1G
A194928-25G
A194928-5G
A194928-250MG
103816-710EA
14
USD
103816-710
103847-324
76785-074
76785-076
77085-266
77085-268
77085-270
77871-970
4-Benzoylbenzoic acid 98%
4-Benzoylbenzoic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₀O₃ Boiling Pt: 415.7 °C (760 mm Hg) Melting Pt: 198…200 °C Flash Pt: 219.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002560 CAS Number: 611-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...