4-(Benzoylamino)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Benzamidobenzoic acid

A101839-250MG A101839-1G
103796-170EA 40 USD
103796-170 103821-864 76809-274 77308-758
4-(Benzoylamino)benzoic acid 95%
4-(Benzoylamino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₃
MW: 241.25 g/mol
Boiling Pt: 349.8 °C (760 mm Hg)
Melting Pt: 288…291 °C
Flash Pt: 165.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00451153
CAS Number: 582-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR