4-Azaindole-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-Pyrrolo[3,2-b]pyridine-3-carboxylic acid
A100273-100MG
A100273-250MG
A100273-5G
103795-336EA
44
USD
103795-336
76780-358
76780-360
76780-362
4-Azaindole-3-carboxylic acid 97%
4-Azaindole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ Boiling Pt: 446.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD11559014 CAS Number: 860496-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...