4-Azaindole 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrolo[3,2-b]pyridine, 1H-Pyrrolo[2,3-e]pyridine

A152186-10G A152186-250MG A152186-1G A152186-25G A152186-5G
103804-354EA 20 USD
103804-354 103842-514 77137-818 77137-820 77318-124 77318-126 77318-128
4-Azaindole 98%
4-Azaindole

Precaución: for Research Use Only

Formula: C₇H₆N₂
Boiling Pt: 273.8 °C (760 mm Hg)
Melting Pt: 126…127 °C
Flash Pt: 124.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00971977
CAS Number: 272-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR