4-Aminotetrahydrofuran-3-ol 95%

Supplier: Ambeed
Warning

A230279-100MG A230279-10G A230279-250MG A230279-1G A230279-5G
103819-534EA 299 USD
103819-534 103850-432 76815-128 76815-130 76815-132 77303-866 77303-868
4-Aminotetrahydrofuran-3-ol 95%
4-Aminotetrahydrofuran-3-ol

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MW: 103.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11870199
CAS Number: 144870-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR