4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine Hydrochloride, 4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride hydrochloride, (1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine hydrochloride
A161694-1G
A161694-5G
103805-520EA
15
USD
103805-520
103843-762
77300-550
77300-552
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride 97%
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO₂S MW: 185.6700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11114559 CAS Number: 116529-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...