5H-Pyrrolo[3,2-d]pyrimidin-4-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Aminopyrrolo[3,2-d]pyrimidine
A172095-100MG
A172095-250MG
A172095-1G
103806-694EA
139
USD
103806-694
76784-270
76784-272
77082-156
5H-Pyrrolo[3,2-d]pyrimidin-4-amine 98%
5H-Pyrrolo[3,2-d]pyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₆H₆N₄ MW: 134.14 g/mol Boiling Pt: 399.8 °C (760 mm Hg) Flash Pt: 224.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11520862 CAS Number: 2227-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...