4-Aminopyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A308784-5G A308784-10G
103814-578EA 11 USD
103814-578 103835-756 76838-518 77015-116
4-Aminopyrimidine-5-carbonitrile 95%
4-Aminopyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₄
MW: 120.11 g/mol
Boiling Pt: 355.8 °C (760 mm Hg)
Melting Pt: 251…255 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00054552
CAS Number: 16357-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR