4-Amino-3-hydroxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-aminopyridine, 4-Aminopyridin-3-ol

A170692-250MG A170692-10G
103806-528EA 85 USD
103806-528 103844-802 77567-504 77671-424
4-Amino-3-hydroxypyridine 97%
4-Amino-3-hydroxypyridine

Precaución: For research use only. Keep in a dark place, inert atmosphere, room temperature

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 447.4 °C (760 mm Hg)
Melting Pt: 222…223 °C
Flash Pt: 224.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01941262
CAS Number: 52334-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR