4-Amino-2-hydroxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-pyridone, 4-Aminopyridin-2(1H)-one, 4-Amino-2(1H)-pyridinone, 4-Amino-2-pyridinol, 4-Amino-pyridin-2-ol, 4-amino-1,2-dihydropyridin-2-one, 3-Deazacytosine, 4-Amino-1H-pyridin-2-one

A265577-100G A265577-10G A265577-25G A265577-5G A265577-1G
103812-738EA 10 USD
103812-738 103852-632 76816-092 76816-094 76816-096 76816-098 77092-332 77304-918
4-Amino-2-hydroxypyridine 97%
4-Amino-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Flash Pt: 141.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02682913
CAS Number: 38767-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR