4-Aminopyridine-2-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A137934-1G A137934-5G A137934-10G
103802-532EA 9 USD
103802-532 77315-528 77315-530 77883-113
4-Aminopyridine-2-carboxylic acid hydrochloride 95%
4-Aminopyridine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18837584
CAS Number: 1291487-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR