4-Aminophthalic acid 95%

Supplier: Ambeed
Warning

A231534-1G A231534-25G A231534-5G A231534-100G
103810-780EA 5 USD
103810-780 103850-530 77089-416 77089-418 77089-420 77113-012
4-Aminophthalic acid 95%
4-Aminophthalic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 465.1 °C (760 mm Hg)
Flash Pt: 235.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00013985
CAS Number: 5434-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR