Fluoral-P (=4-amino-3-penten-2-one) 96%

Supplier: Ambeed
Warning

A157917-10G A157917-1G A157917-5G
103805-062EA 7 USD
103805-062 103843-268 77286-340 77300-300 77300-302
Fluoral-P (=4-amino-3-penten-2-one) 96%
Fluoral-P (=4-amino-3-penten-2-one)

Precaución: for Research Use Only

Formula: C₅H₉NO
Boiling Pt: 130…131 °C (760 mm Hg)
Melting Pt: 38 °C
Storage Temperature: Freezer
MDL Number: MFCD00043715
CAS Number: 1118-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR