4-Aminonicotinamide 98%

Supplier: Ambeed
Warning

A141785-100MG A141785-250MG A141785-25G A141785-1G A141785-5G
103803-058EA 245 USD
103803-058 76871-250 76871-252 76871-254 77316-246 77316-248
4-Aminonicotinamide 98%
4-Aminonicotinamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
MW: 137.1400 g/mol
Boiling Pt: 377.8 °C (760 mm Hg)
Melting Pt: 235…236 °C
Storage Temperature: Ambient
MDL Number: MFCD09263484
CAS Number: 7418-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR