4-Amino-N,N-dimethylbenzamide 97%
Supplier: Ambeed
|
Warning
|
A101804-100MG
A101804-10G
A101804-250MG
A101804-25G
A101804-1G
A101804-5G
103796-146EA
20
USD
103796-146
103821-834
77245-146
77245-148
77245-150
77245-152
77308-732
77308-734
4-Amino-N,N-dimethylbenzamide 97%
4-Amino-N,N-dimethylbenzamide
Precaución: for Research Use Only
Formula:
C₉H₁₂N₂O Boiling Pt: 348 °C (760 mm Hg) Melting Pt: 151…154 °C Flash Pt: 164.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00523648 CAS Number: 6331-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...