4-Amino-N,N,3-trimethylbenzamide 95%

Supplier: Ambeed
Warning

A103019-1G A103019-5G A103019-1G
77309-034EA 41 USD
77309-034 103822-168 103796-466
4-Amino-N,N,3-trimethylbenzamide 95%
4-Amino-N,N,3-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09728471
CAS Number: 953739-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR