4-Benzylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Aminodiphenylmethane

A111827-1G A111827-5G
103797-908EA 5 USD
103797-908 103823-730 77310-538 77310-540
4-Benzylaniline 98%
4-Benzylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.25 g/mol
Boiling Pt: 142…144 °C (760 mm Hg)
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00047861
CAS Number: 1135-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR