4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) 95%

Supplier: Ambeed
Warning

Synonyms: 4-Aminocyclohexanecarboxylic acid, 4-Aminocyclohexane-1-carboxylic acid

A100757-100G A100757-10G A100757-250MG A100757-1G A100757-25G
103795-610EA 14 USD
103795-610 103821-306 77107-166 77107-168 77107-170 77282-556 77684-925
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers) 95%
4-Aminocyclohexanecarboxylic acid (mixture of cis and trans isomers)

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.1800 g/mol
Melting Pt: 299…301 °C
Flash Pt: 123.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00191601
CAS Number: 1776-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR