4-Aminobutyric acid 98%
Supplier: Ambeed
|
Warning
|
A246764-1000G
A246764-100G
A246764-10G
A246764-500G
A246764-25G
A246764-5G
103851-564EA
5
USD
103851-564
76976-588
76976-590
76976-592
76976-594
77090-866
77090-868
4-Aminobutyric acid 98%
4-Aminobutyric acid
Precaución: for Research Use Only
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 248 °C (760 mm Hg) Melting Pt: 195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...