4-Aminobutanamide hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A403939-1G
A403939-250MG
103827-098EA
52
USD
103827-098
76841-616
77672-974
4-Aminobutanamide hydrochloride 95%
4-Aminobutanamide hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₁ClN₂O MW: 138.59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792526 CAS Number: 13031-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...