4-Aminobicyclo[2.2.2]octan-1-ol hydrochloride 97%

Supplier: Ambeed
Warning

A100144-100MG A100144-250MG A100144-50MG A100144-1G
103795-270EA 469 USD
103795-270 76982-114 76982-116 76982-118 77296-578
4-Aminobicyclo[2.2.2]octan-1-ol hydrochloride 97%
4-Aminobicyclo[2.2.2]octan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO
MW: 177.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26406403
CAS Number: 1403864-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR