4-Aminobenzamidine dihydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Aminophenylamidine dihydrochloride, 4-Aminobenzene-1-carboximidamide dihydrochloride
A111482-10G
A111482-250MG
A111482-1G
103797-844EA
14
USD
103797-844
103823-664
77270-924
77270-926
77310-466
4-Aminobenzamidine dihydrochloride 95%
4-Aminobenzamidine dihydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₁Cl₂N₃ MW: 208.09 g/mol Melting Pt: >300 °C Flash Pt: 129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013001 CAS Number: 2498-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...