4-Amino-6-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A108326-10G A108326-25G A108326-5G
103823-106EA 5 USD
103823-106 77008-772 77008-774 77283-708
4-Amino-6-methoxypyrimidine 98%
4-Amino-6-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.1286 g/mol
Boiling Pt: 290.8 °C (760 mm Hg)
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00129983
CAS Number: 696-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR