4-Amino-6-chloropyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

A170023-250MG A170023-1G A170023-5G A170023-10G
103806-438EA 22 USD
103806-438 103844-704 76784-170 77081-802 77081-804 77667-380
4-Amino-6-chloropyrimidine-5-carbonitrile 97%
4-Amino-6-chloropyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MW: 154.5600 g/mol
Boiling Pt: 402.4 °C (760 mm Hg)
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07761812
CAS Number: 60025-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR