4-Amino-6-chloro-3-pyridinecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-chloronicotinic acid

A164727-100MG A164727-10G A164727-250MG A164727-1G
103805-894EA 33 USD
103805-894 103844-152 76872-158 76872-160 76872-162 77320-338
4-Amino-6-chloro-3-pyridinecarboxylic acid 95%
4-Amino-6-chloro-3-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.569 g/mol
Boiling Pt: 433.9±45.0 °C (760 mm Hg)
Flash Pt: 216.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234161
CAS Number: 1060811-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR