4-Amino-6-chloronicotinaldehyde 97%

Supplier: Ambeed
Warning

A247014-1G A247014-5G A247014-250MG A247014-25G
103811-744EA 25 USD
103811-744 103851-578 77090-882 77090-884 77672-202 77883-663
4-Amino-6-chloronicotinaldehyde 97%
4-Amino-6-chloronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O
Boiling Pt: 379 °C (760 mm Hg)
Flash Pt: 183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834660
CAS Number: 1001756-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR