4-Amino-6-chloro-2-(methylthio)pyrimidine 95%

Supplier: Ambeed
Warning

A132073-10G A132073-25G A132073-1G A132073-5G A132073-100G
103801-686EA 13 USD
103801-686 77010-002 77298-534 77314-242 77314-244 77683-171
4-Amino-6-chloro-2-(methylthio)pyrimidine 95%
4-Amino-6-chloro-2-(methylthio)pyrimidine

Precaución: For research use only.

Formula: C₅H₆ClN₃S
MW: 175.6400 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00006088
CAS Number: 1005-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR