4-Amino-6-bromoquinoline 98%

Supplier: Ambeed
Warning

A112680-100MG A112680-10G A112680-250MG A112680-1G A112680-5G
103798-072EA 79 USD
103798-072 103823-910 77160-824 77160-826 77160-828 77310-720 77310-722
4-Amino-6-bromoquinoline 98%
4-Amino-6-bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
Boiling Pt: 388.7 °C (760 mm Hg)
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD03407953
CAS Number: 65340-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR